Lead(II) acetate basic, anhydrous, EMSURE® for the analysis of sugar acc. to horne, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.07414.1000
1.07414.9030
CA1.07414.1000EA
197
CAD
CA1.07414.1000
CA1.07414.9030
Lead(II) acetate basic, anhydrous, EMSURE® for the analysis of sugar acc. to horne, Supelco®
Lead (II) acetate basic
Formula:
(CH₃COO)₂Pb·2Pb(OH)₂ Melting Pt: 198…205 °C Density: 3.58 g/cm3 (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011154 CAS Number: 1335-32-6 EINECS: 215-630-3 UN: 3077 ADR: 9,III |
Specification Test Results
Assay (PbO) | ≥ 33.0 % |
In water insoluble matter | ≤ 1.0 % |
Substances insoluble in dilute acetic acid | ≤ 0.02 % |
Chloride (Cl) | ≤ 0.003 % |
Nitrate und Nitrite (as NO₃) | passes test |
Ca (Calcium) | ≤ 0.01 % |
Cu (Copper) | ≤ 0.002 % |
Fe (Iron) | ≤ 0.002 % |
K (Potassium) | ≤ 0.02 % |
Na (Sodium) | ≤ 0.05 % |
Loss on Drying (105 °C) | ≤ 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...