Nickel (II) sulfate hexahydrate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Nickelous sulphate, Hexahydrate, Nickelous sulfate, Hexahydrate, Nickel sulfate hexahydrate, Nickel sulphate hexahydrate

1.06727.0100 1.06727.0250 1.06727.1000
CA1.06727.0100EA 169 CAD
CA1.06727.0100 CA1.06727.0250 CA1.06727.1000
Nickel (II) sulfate hexahydrate, EMSURE® ACS for analysis, Supelco®
Nickel sulphate hexahydrate
Formula: NiSO₄·6H₂O
Melting Pt: 840 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3077
ADR: 9,III
Merck Index: 13,06541

Order Now

Specification Test Results

Solubility 650 g/l (20 °C)
Melting Point 53 °C
Molar Mass 262.86 g/mol
Bulk Density 1000 kg/m3
Density 2.07 g/cm3 (20 °C)
pH 4.3 - 4.7 (100 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR