Nickel(II) nitrate hexahydrate, EMSURE® ACS analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

1.06721.0100 1.06721.0250 1.06721.1000
CA1.06721.0100EA 120 CAD
CA1.06721.0100 CA1.06721.0250 CA1.06721.1000
Nickel(II) nitrate hexahydrate, EMSURE® ACS analysis_for, Supelco®
Nickel (II) nitrate hexahydrate
Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137
Melting Pt: 56.7
Density: 2.05 g/cm³ (20 °C)
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1 III
Merck Index: 12,06596

Order Now

Specification Test Results

Solubility 940 g/l (20 °C)
Melting Point 56.7 °C
Molar Mass 290.81 g/mol
Bulk Density 800 kg/m3
Boiling Point 136.7 °C (1013 hPa)
Density 2.05 g/cm3 (20 °C)
pH 5 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR