Nickel(II) chloride hexahydrate, EMSURE® ACS for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.06717.0250
1.06717.1000
CA1.06717.0250EA
188
CAD
CA1.06717.0250
CA1.06717.1000
Nickel(II) chloride hexahydrate, EMSURE® ACS for analysis, Supelco®
Nickel (II) chloride hexahydrate
Formula:
NiCl₂·6H₂O Melting Pt: 140 °C Density: 1.84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149809 CAS Number: 7791-20-0 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 13,06529 |
Specification Test Results
Solubility | 553 g/l (20 °C) |
Melting Point | 1001 °C (anhydrous substance),decomposes |
Molar Mass | 237.66 g/mol |
Bulk Density | 640 kg/m3 |
Boiling Point | 987 °C (sublimed),(anhydrous substance) |
pH | 4.9 (100 g/l, H2O, 20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...