Sodium dichromate dihydrate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06336.0250 1.06336.1000
CA1.06336.0250EA 125 CAD
CA1.06336.0250 CA1.06336.1000
Sodium dichromate dihydrate, EMSURE® ACS for analysis, Supelco®
Sodium dichromate dihydrate
Formula: Na₂Cr₂O₇·2H₂O
MW: 298.00 g/mol (261.97 anhy)
Melting Pt: 356.7
Density: 2.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3086
ADR: 6.1 (5.1) II
Merck Index: 12,08754

Order Now

Specification Test Results

Solubility 731.8 g/l (20 °C)
Melting Point 356.7 °C (anhydrous substance)
Molar Mass 298.00 g/mol
Bulk Density 1200 kg/m3
Boiling Point 400 °C (decomposition)
Density 2.35 g/cm3 (20 °C)
pH 3.5 (100 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR