Methanol 99,9 GC_ASSAY_METHOD, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

1.06009.1011 1.06009.2500 1.06009.2511 1.06009.5000 1.06009.6010 1.06009.6025 1.06009.9025 1.06009.9180 1.06009.1000
CA1.06009.1011CS 427 CAD
CA1.06009.1011 CA1.06009.2500 CA1.06009.2511 CA1.06009.5000 CA1.06009.6010 CA1.06009.6025 CA1.06009.9025 CA1.06009.9180 CA1.06009.1000
Methanol 99,9 GC_ASSAY_METHOD, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 (lit.)
Melting Pt: -98 (lit.)
Density: 0.791 g/ml (25°C) (lit.)
Flash Pt: 9.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,6.1 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Appearance clear
Color ≤ 10 Hazen
Solubility in water conforms
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Density (d 20 °C/20 °C) 0.791 - 0.793
Boiling point 64 - 65 °C
Benzene (impurity A) (GC) ≤ 2 ppm
Ethanol (GC) ≤ 0.05 %
Acetone ≤ 0.001 %
Acetaldehyde ≤ 0.001 %
Formaldehyde ≤ 0.001 %
Readily carbonizable substances conforms
Carbonyl compounds (as CO) ≤ 0.001 %
Chloride (Cl) ≤ 0.5 ppm
Sulfate (SO₄) ≤ 1 ppm
Substances reducing potassium permanganate (as O) ≤ 0.00025 %
Ag (Silver) ≤ 0.000002 %
Al (Aluminium) ≤ 0.00005 %
As (Arsenic) ≤ 0.000002 %
Au (Gold) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Be (Beryllium) ≤ 0.000002 %
Bi (Bismuth) ≤ 0.000002 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Ga (Gallium) ≤ 0.000002 %
In (Indium) ≤ 0.000002 %
Li (Lithium) ≤ 0.000002 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Mo (Molybdenum) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Pt (Platinum) ≤ 0.000005 %
Sn (Tin) ≤ 0.00001 %
Ti (Titanium) ≤ 0.000002 %
Tl (Thallium) ≤ 0.000002 %
V (Vanadium) ≤ 0.000002 %
Zn (Zinc) ≤ 0.00001 %
Zr (Zirconium) ≤ 0.000002 %
Evaporation residue ≤ 0.0005 %
Water ≤ 0.05 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR