Manganese(II) sulfate monohydrate, spray-dried 99.0-101.0% (by complexometry from ignited basis) [98.0-101.0% (MnSO4.H2O basis, complexometric)], EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate
1.05941.0250
1.05941.9025
CA1.05941.0250EA
193
CAD
CA1.05941.0250
CA1.05941.9025
Manganese(II) sulfate monohydrate, spray-dried 99.0-101.0% (by complexometry from ignited basis) [98.0-101.0% (MnSO4.H2O basis, complexometric)], EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Manganese (II) sulphate monohydrate
1.05941.0250 is offered as an alternative to MX0210-1. Different Pack Size, Same Specifications.
1.05941.9025 is offered as an alternative to MX0210-30. Different Pack Size, Same Specifications.
Formula:
MnSO₄·1H₂O MW: 169.02 g/mol Melting Pt: >449 °C (anhydr.) Density: 2.95 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149159 CAS Number: 10034-96-5 EINECS: 232-089-9 UN: 3077 ADR: 9 III Merck Index: 13,05763 |
Specification Test Results
Solubility | 762 g/l (20 °C) |
Melting Point | 700 °C (anhydrous substance) |
Molar Mass | 169.02 g/mol |
Bulk Density | 1000 - 1200 kg/m3 |
Density | 2.95 g/cm3 (20 °C) |
pH | 3.0 - 3.5 (50 g/l, H2O, 20 °C) |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous hazardous material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...