Mercury(II) iodide, red Reag. Ph. Eur., ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Mercury diiodide, Mercuric iodide

M-Clarity™ quality level = MQ300

1.04428.0050 1.04428.0250
CA1.04428.0050EA 347 CAD
CA1.04428.0050 CA1.04428.0250
Mercury(II) iodide, red Reag. Ph. Eur., ACS for analysis, Supelco®
Mercury (II) iodide
Formula: HgI₂
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 259 °C
Density: 6.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011044
CAS Number: 7774-29-0
EINECS: 231-873-8
UN: 1638
ADR: 6.1,II
Merck Index: 13,05905

Order Now

Specification Test Results

Assay (argentometric, dried sust.) ≥ 99.0 %
Solubility in Potassium iodide passes test
Soluble mercury salts (as Hg) ≤ 0.05 %
Foreign heavy metals (as Pb) ≤ 0.001 %
Fe (Iron) ≤ 0.001 %
Mercury (I) (as Hg) ≤ 0.1 %
Substances not reducing by formic acid ≤ 0.02 %
Corresponds to ACS,Reag. Ph Eur


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR