Mercury(II) bromide ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.04421.0050
1.04421.0250
CA1.04421.0050EA
315
CAD
CA1.04421.0050
CA1.04421.0250
Mercury(II) bromide ACS, Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) bromide
Formula:
Br₂Hg Boiling Pt: 320 °C (1013 hPa) Melting Pt: 236 °C Density: 6.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011040 CAS Number: 7789-47-1 EINECS: 232-169-3 UN: 1634 ADR: 6.1,II Merck Index: 13,05900 |
Specification Test Results
Assay (complexometric) | ≥ 99.0 % |
Insoluble matter (in methanol) | ≤ 0.05 % |
Chloride (Cl) | ≤ 0.25 % |
Residue after reduction | ≤ 0.02 % |
Correspons to ACS,Reag. Ph Eur |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...