Mercury(II) chloride ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.04419.0050
1.04419.0250
1.04419.1000
CA1.04419.0050EA
284
CAD
CA1.04419.0050
CA1.04419.0250
CA1.04419.1000
Mercury(II) chloride ACS, Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) chloride
Formula:
HgCl₂ Boiling Pt: 302 °C (1013 hPa) Melting Pt: 273 °C Density: 5.44 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011041 CAS Number: 7487-94-7 EINECS: 231-299-8 UN: 1624 ADR: 6.1,II Merck Index: 13,05901 |
Specification Test Results
Assay (complexometric) | ≥ 99.5 % |
Assay (complexometric; calculated on dried substance) | 99.5 - 100.5 % |
Identity | passes test |
Appearance of solution | passes test |
Solution in Ether | passes test |
Acidity or alkalinity | passes test |
Mercury(I)-chloride | passes test |
Fe (Iron) | ≤ 0.002 % |
Residue after reduction | ≤ 0.02 % |
Loss on drying (24 h im Vakuum) | ≤ 1.0 % |
Correspons to Reag. Ph Eur,ACS |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...