Mercury(II) acetate ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

1.04410.0050 1.04410.0250
CA1.04410.0050EA 222 CAD
CA1.04410.0050 CA1.04410.0250
Mercury(II) acetate ACS, Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
MW: 318.68 g/mol
Melting Pt: 178…180 °C
Density: 3.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1 II
Merck Index: 13,05898

Order Now

Specification Test Results

Solubility 400 g/l (20 °C)
Melting Point 178 - 180 °C (decomposition)
Molar Mass 318.68 g/mol
Bulk Density 1000 kg/m3
Density 3.27 g/cm3 (20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR