Dimethyl sulfoxide ≥99.9% (by GC), EMSURE® ACS for analysis, Supelco®
Supplier: MilliporeSigma
1.02952.1000
1.02952.1011
1.02952.2500
1.02952.2511
1.02952.9025
CA1.02952.1000CS
1796
CAD
CA1.02952.1000
CA1.02952.1011
CA1.02952.2500
CA1.02952.2511
CA1.02952.9025
Dimethyl sulfoxide ≥99.9% (by GC), EMSURE® ACS for analysis, Supelco®
Dimethyl sulphoxide
Formula:
(CH₃)₂SO Boiling Pt: 189 °C (lit.) Melting Pt: 16…19 °C (lit.) Density: 1.10 g/ml (lit.) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
|
Specification Test Results
Solubility | 1000 g/l (20 °C) |
Melting Point | 18.4 °C |
Molar Mass | 78.13 g/mol |
Boiling Point | 189 °C (1013 hPa) |
Vapor Pressure | 0.6 hPa (20 °C) |
Flash Point | 87 °C |
Refractive Index | 1.48 (20 °C) |
Explosion Limit | 1.8 - 63.0 %(V) |
Density | 1.10 g/cm3 (20 °C) |
Ignition Point | 300 - 302 °C |
Saturation concentration | 8.0 g/m3 (20 °C) |
Viscosity kinematic | 2.14 mm2/s |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...