Celite® 545, filter aid 0.02-0.1 mm, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: DE, Diatomaceous earth, flux-calcined

1.02693.0250 1.02693.1000 1.02693.9010 1.02693.0250 1.02693.1000
89501-602EA 196.65 CAD
89501-602 89501-604 89501-606 CA89501-602 CA89501-604
Celite® 545, filter aid 0.02-0.1 mm, Supelco®
Celite® 545, filter aid
Formula: SiO₂
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 272-489-0

Order Now

Specification Test Results

Solubility  - 0.001 g/l (20 °C)
Bulk Density 300 kg/m3
pH 10 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR