Celite® 281 pH 10 (100 g/l, H₂O, 20 °C), Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.02686.1000
1.02686.1000
CA89501-598EA
444.04
CAD
CA89501-598
89501-598
Celite® 281 pH 10 (100 g/l, H₂O, 20 °C), Supelco®
Celite® 281
CAS Number:
68855-54-9 |
Specification Test Results
Solubility |
|
Bulk Density | 300 kg/m3 |
pH | 10 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...