1-Butanol ≥99.5% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

1.01990.1000 1.01990.2500 1.01990.6025
CA1.01990.1000CS 974 CAD
CA1.01990.1000 CA1.01990.2500 CA1.01990.6025
1-Butanol ≥99.5% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 116…118 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.81 g/cm3 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 77 g/l (20 °C)
Melting Point -89 °C
Molar Mass 74.12 g/mol
Boiling Point 116 - 118 °C (1013 hPa)
Vapor Pressure 6.7 hPa (20 °C)
Flash Point 34 °C
Refractive Index 1.3993 (20 °C, 589 nm)
Explosion Limit 1.4 - 11.3 %(V)
Density 0.81 g/cm3 (20 °C)
pH 7 (70 g/l, H2O, 20 °C)
Ignition Point 340 °C
Evaporation number 33

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR