tert-Butyl methyl ether, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

1.01849.1000 1.01849.2500
CA1.01849.1000CS 1113 CAD
CA1.01849.1000 CA1.01849.2500
tert-Butyl methyl ether, EMSURE® for analysis, Supelco®
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2
Melting Pt: -108.6
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Color ≤ 10 Hazen
Acidity ≤ 0.0005 meq/g
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %
Peroxide (as H₂O₂) ≤ 0.0001 %
Evaporation residue ≤ 0.001 %
Water ≤ 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR