Petroleum spirit, 30…50 °C, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

1.01786.1000 1.01786.2500
CA1.01786.1000CS 2422 CAD
CA1.01786.1000 CA1.01786.2500
Petroleum spirit, 30…50 °C, EMSURE® for analysis, Supelco®
Petroleum spirit
Boiling Pt: 30…50 °C (1013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,I

Order Now

Specification Test Results

Solubility (25 °C) insoluble
Boiling Point 30 - 50 °C (1013 hPa)
Explosion Limit 1.2 - 7.5 %(V)
Density 0.65 g/cm3 (20 °C)
Ignition Point 285 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR