Petroleum spirit, 80…100 °C, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Petroleum etherGasolinePetrolBenzine

1.01777.1000 1.01777.1000
CA1.01777.1000CS 1476.45 CAD
EM1.01777.1000 CA1.01777.1000
Petroleum spirit, 80…100 °C, EMSURE® for analysis, Supelco®
Petroleum spirit
Boiling Pt: 80…100 °C (1013 hPa)
Density: 0.69 g/cm³ (20 °C)
Flash Pt: –10 °C
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Solubility (25 °C) insoluble
Boiling Point 80 - 100 °C (1013 hPa)
Vapor Pressure 60 hPa (20 °C)
Explosion Limit 1.0 - 6.5 %(V)
Density 0.69 g/cm3 (20 °C)
Ignition Point 250 °C
Viscosity kinematic 0.6 mm2/s (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR