Petroleum spirit, 40…60 °C, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

1.01775.1000 1.01775.2500 1.01775.6010 1.01775.6025
CA1.01775.1000CS 980 CAD
CA1.01775.1000 CA1.01775.2500 CA1.01775.5000 CA1.01775.6010 CA1.01775.6025
Petroleum spirit, 40…60 °C, EMSURE® ACS for analysis, Supelco®
Petroleum spirit
Boiling Pt: 36…83 °C (1013 hPa)
Melting Pt: <–80 °C
Density: 0.645…0.665 g/cm3 (15 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Boiling Point 36 - 83 °C (1013 hPa)
Vapor Pressure 370 hPa (25 °C)
Explosion Limit 1.0 - 7.4 %(V)
Density 0.645 - 0.665 g/cm3 (15 °C)
pH (H2O) not applicable
Viscosity kinematic 0.45 mm2/s (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR