Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

1.01774.1000 1.01774.2500 1.01774.6025
CA1.01774.1000CS 1288 CAD
CA1.01774.1000 CA1.01774.2500 CA1.01774.5000 CA1.01774.6025
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Petroleum spirit
Boiling Pt: 60…80 °C (1013 hPa)
Density: 0.68 g/cm3 (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Solubility (25 °C) practically insoluble
Boiling Point 60 - 80 °C
Vapor Pressure 200 hPa (20 °C)
Flash Point -26 °C
Explosion Limit 1.0 - 7.4 %(V)
Density 0.68 g/cm3 (20 °C)
Ignition Point 240 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR