1-Propanol ≥99.5% (by GC), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00997.1000 1.00997.2500 1.00997.6025
CA1.00997.1000CS 1252 CAD
CA1.00997.1000 CA1.00997.2500 CA1.00997.6025
1-Propanol ≥99.5% (by GC), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
1-Propanol
Formula: H₃CCH₂CH₂OH
Boiling Pt: 96.5…98 °C (1013 hPa)
Melting Pt: –127 °C
Density: 0.80 g/cm3 (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) miscible in all proportions
Melting Point -127 °C
Molar Mass 60.1 g/mol
Boiling Point 96.5 - 98 °C (1013 hPa)
Vapor Pressure 19 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 2.1 - 19.2 %(V)
Density 0.80 g/cm3 (20 °C)
pH 7 (200 g/l, H2O, 20 °C)
Ignition Point 360 °C
Evaporation number 17


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR