Isobutanol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.00984.1000
1.00984.2500
CA1.00984.1000CS
1107
CAD
CA1.00984.1000
CA1.00984.2500
Isobutanol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Isobutanol
Formula:
(CH₃)₂CHCH₂OH Boiling Pt: 107.9 °C (1013 hPa) Melting Pt: –108 °C Density: 0.802 g/cm³ (20 °C) Flash Pt: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004740 CAS Number: 78-83-1 EINECS: 201-148-0 UN: 1212 ADR: 3,III Merck Index: 13,05148 |
|
Specification Test Results
Solubility | 85 g/l (20 °C) |
Melting Point | -108 °C |
Molar Mass | 74.12 g/mol |
Boiling Point | 106 - 108 °C (1013 hPa) |
Vapor Pressure | 12 hPa (20 °C) |
Flash Point | 28 °C |
Explosion Limit | 1.5 - 12 %(V) |
Density | 0.802 g/cm3 (20 °C) |
pH | 7 (80 g/l, H2O, 20 °C) |
Ignition Point | 430 °C |
Evaporation number | 24 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...