Isoamyl alcohol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.00979.1000
1.00979.2500
CA1.00979.1000CS
1747
CAD
CA1.00979.1000
CA1.00979.2500
Isoamyl alcohol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Isoamyl alcohol
Formula:
(CH₃)₂CHCH₂CH₂OH Boiling Pt: 130 °C (1013 hPa) Melting Pt: –117 °C Density: 0.812 g/cm³ (20 °C) Flash Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,III Merck Index: 13,05214 |
|
Specification Test Results
Solubility | 25 g/l (20 °C) |
Melting Point | -117 °C |
Molar Mass | 88.15 g/mol |
Boiling Point | 131 °C (1013 hPa) |
Vapor Pressure | 3 hPa (20 °C) |
Flash Point | 45 °C |
Explosion Limit | 1.2 - 8 %(V) |
Density | 0.81 g/cm3 (20 °C) |
pH | 7 (25 g/l, H2O, 20 °C) |
Ignition Point | 335 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...