Ethanol denatured ≥99.5% (by GC), EMSURE® for analysis (denatured with 1% MEK), Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Ethyl alcohol denatured
M-Clarity™ quality level = MQ300
1.00974.1011
1.00974.6025
1.00974.9025
1.00974.9180
EM1.00974.1011CS
1027
CAD
EM1.00974.1011
CA1.00974.6025
CA1.00974.9025
CA1.00974.9180
CA1.00974.2500
Ethanol denatured ≥99.5% (by GC), EMSURE® for analysis (denatured with 1% MEK), Supelco®
Ethanol denatured
Formula:
H₃CCH₂OH MW: 46.07 g/mol Melting Pt: –114.5 °C (ethanol) Density: 0.79 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3 II |
Specification Test Results
Purity (GC) | ≥ 99.5 % |
Identity (IR) | conforms |
Free acid (as acetic acid) | ≤ 0.001 % |
Ethylmethylketone (GC) | 0.8 - 1.3 % |
Readily carbonizable substances | conforms |
Evaporation residue | ≤ 0.001 % |
Water | ≤ 0.1 % |
Al (Aluminium) | ≤ 0.00005 % |
B (Boron) | ≤ 0.00005 % |
Ba (Barium) | ≤ 0.00001 % |
Ca (Calcium) | ≤ 0.00005 % |
Cd (Cadmium) | ≤ 0.000005 % |
Co (Cobalt) | ≤ 0.000002 % |
Cr (Chromium) | ≤ 0.000002 % |
Cu (Copper) | ≤ 0.000002 % |
Fe (Iron) | ≤ 0.00001 % |
K (Potassium) | ≤ 0.0001 % |
Mg (Magnesium) | ≤ 0.00001 % |
Mn (Manganese) | ≤ 0.000002 % |
Ni (Nickel) | ≤ 0.000002 % |
Pb (Lead) | ≤ 0.00001 % |
Sn (Tin) | ≤ 0.00001 % |
Zn (Zinc) | ≤ 0.00001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...