Ethanolamine, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00845.1000 1.00845.2500
CA1.00845.1000CS 2493 CAD
CA1.00845.1000 CA1.00845.2500
Ethanolamine, EMSURE® for analysis, Supelco®
Ethanolamine
Formula: NH₂CH₂CH₂OH
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: 10.5 °C
Density: 1.0164 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8,III
Merck Index: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 10.5 °C
Molar Mass 61.08 g/mol
Boiling Point 171 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 92.5 °C
Refractive Index 1.4539
Explosion Limit 3.4 - 27 %(V)
Density 1.02 g/cm3 (20 °C)
pH 12.1 (100 g/l, H2O, 20 °C)
Ignition Point 410 °C
Viscosity kinematic 20 mm2/s (23 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR