Trichloroacetic acid (TCA), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00807.0100 1.00807.1000 1.00807.0250
CA1.00807.0100CS 726 CAD
CA1.00807.0100 CA1.00807.1000 CA1.00807.0250
Trichloroacetic acid (TCA), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 1839
ADR: 8,II
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1600 g/l (20 °C)
Melting Point 54 - 56 °C
Molar Mass 163.38 g/mol
Bulk Density 900 kg/m3
Boiling Point 197 °C
Vapor Pressure 1 hPa (20 °C)
Flash Point 110 °C
Density 1.63 g/cm3 (20 °C)
pH  - 1 (50 g/l, H2O, 20 °C)
Ignition Point 711 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR