Sulfurous acid (5 - 6% SO₂) in aqueous solution, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.00761.1000
1.00761.2500
CA1.00761.1000CS
835
CAD
CA1.00761.1000
CA1.00761.2500
Sulfurous acid (5 - 6% SO₂) in aqueous solution, EMSURE® for analysis, Supelco®
Sulfurous acid
Formula:
H₂O₃S Density: 1.03 g/cm³ (20 °C) |
MDL Number:
MFCD00011335 CAS Number: 7782-99-2 EINECS: 231-973-1 UN: 1833 ADR: 8,II |
Specification Test Results
Density | 1.03 g/cm3 (20 °C) |
pH | (H2O, 20 °C) acidic |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...