Sulfuric acid 95.0-97.0% (by alkalimetry), EMSURE® ISO for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.00731.1000
1.00731.1011
1.00731.2500
1.00731.2510
1.00731.2511
1.00731.9025
CA1.00731.1000CS
565
CAD
CA1.00731.1000
CA1.00731.1011
CA1.00731.2500
CA1.00731.2510
CA1.00731.2511
CA1.00731.9025
Sulfuric acid 95.0-97.0% (by alkalimetry), EMSURE® ISO for analysis, Supelco®
Sulfuric acid
Packaging: Safebreak is a robust glass bottle coated with polyethylene (PE). Should the bottle fall and break, all acid and glass splinters are safely retained within the polyethylene coating, thus protecting you from any risk.
Formula:
H₂SO₄ Boiling Pt: ~290 °C (lit.) Melting Pt: –20 °C Density: 1.840 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II Merck Index: 14,08974 |
Specification Test Results
Assay (alkalimetric) | 95.0 - 97.0 % |
Color | ≤ 10 Hazen |
Chloride (Cl) | ≤ 0.1 ppm |
Nitrate (NO₃) | ≤ 0.2 ppm |
Phosphate (PO₄) | ≤ 0.5 ppm |
Ag (Silver) | ≤ 0.010 ppm |
Al (Aluminium) | ≤ 0.050 ppm |
As (Arsenic) | ≤ 0.010 ppm |
Au (Gold) | ≤ 0.050 ppm |
B (Boron) | ≤ 0.050 ppm |
Ba (Barium) | ≤ 0.050 ppm |
Be (Beryllium) | ≤ 0.010 ppm |
Bi (Bismuth) | ≤ 0.050 ppm |
Ca (Calcium) | ≤ 0.100 ppm |
Cd (Cadmium) | ≤ 0.010 ppm |
Co (Cobalt) | ≤ 0.010 ppm |
Cr (Chromium) | ≤ 0.020 ppm |
Cu (Copper) | ≤ 0.010 ppm |
Fe (Iron) | ≤ 0.100 ppm |
Ga (Gallium) | ≤ 0.050 ppm |
Ge (Germanium) | ≤ 0.020 ppm |
In (Indium) | ≤ 0.050 ppm |
K (Potassium) | ≤ 0.100 ppm |
Li (Lithium) | ≤ 0.010 ppm |
Mg (Magnesium) | ≤ 0.050 ppm |
Mn (Manganese) | ≤ 0.010 ppm |
Mo (Molybdenum) | ≤ 0.020 ppm |
N/A (Sodium) | ≤ 0.300 ppm |
NH₄ (Ammonium) | ≤ 1 ppm |
Ni (Nickel) | ≤ 0.020 ppm |
Pb (Lead) | ≤ 0.010 ppm |
Pt (Platinum) | ≤ 0.100 ppm |
Sn (Tin) | ≤ 0.050 ppm |
Sr (Strontium) | ≤ 0.010 ppm |
Ti (Titanium) | ≤ 0.020 ppm |
Tl (Thallium) | ≤ 0.020 ppm |
V (VaN/Adium) | ≤ 0.010 ppm |
Zn (Zinc) | ≤ 0.050 ppm |
Zr (Zirconium) | ≤ 0.020 ppm |
Substances reducing potassium permangaN/Ate (as SO₂) | ≤ 2 ppm |
Residue on ignition | ≤ 3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...