Phosphotungstic acid hydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00583.0100 1.00583.0250
CA1.00583.0100EA 473 CAD
CA1.00583.0100 CA1.00583.0250
Phosphotungstic acid hydrate, EMSURE® for analysis, Supelco®
Phosphotungstic acid hydrate

1.00583.0100 replaces and is the same as PX1055-4. Part # Change only, Same Pack Size, Same Specifications.

Formula: H₃[P(W₃O₁₀)₄]·nH₂O
Melting Pt: 107 °C
MDL Number: MFCD00011342
CAS Number: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 13,07450

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 107 °C
Molar Mass 2880.17 g/mol (anhydrous)
Bulk Density 960 kg/m3
pH 1.3 (20 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR