Phosphotungstic acid hydrate, crystals, Extra Pure, Supelco®

Supplier: MilliporeSigma
Danger

1.00582.0100 1.00582.9025
CA1.00582.0100EA 590 CAD
CA1.00582.0100 CA1.00582.9025
Phosphotungstic acid hydrate, crystals, Extra Pure, Supelco®
Phosphotungstic acid hydrate
Formula: H₃[P(W₃O₁₀)₄]·nH₂O
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00011342
CAS Number: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 13,07450

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 107 °C
Molar Mass 2880.17 g/mol (anhydrous)
Bulk Density 960 kg/m3
pH 1.3 (20 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR