Orthophosphoric acid ≥85%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.00573.1000 1.00573.2500 1.00573.2510 1.00573.9025
CA1.00573.1000CS 1167 CAD
CA1.00573.1000 CA1.00573.2500 CA1.00573.2510 CA1.00573.9025
Orthophosphoric acid ≥85%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Orthophosphoric acid
Formula: H₃PO₄
Boiling Pt: 158 °C (lit.)
Melting Pt: ~40 °C (lit.)
Density: 1.685 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Assay (alkalimetric) ≥ 85.0 %
Identity passes test
Appearance of solution passes test
Color ≤ 10 Hazen
Chloride (Cl) ≤ 2 ppm
Fluoride (F) ≤ 1 ppm
Nitrate (NO₃) ≤ 3 ppm
Phosphite and Hypophosphite (as H₃PO₃) ≤ 20 ppm
Hypophosphite, phosphite passes test
Heavy metals (as Pb) ≤ 10 ppm
Sulfate (SO₄) ≤ 20 ppm
As (Arsenic) ≤ 0.5 ppm
Ca (Calcium) ≤ 20 ppm
Cd (Cadmium) ≤ 0.5 ppm
Co (Cobalt) ≤ 0.5 ppm
Cu (Copper) ≤ 0.5 ppm
Fe (Iron) ≤ 10 ppm
K (Potassium) ≤ 5 ppm
Mg (Magnesium) ≤ 5 ppm
Mn (Manganese) ≤ 0.5 ppm
Na (Sodium) ≤ 200 ppm
Ni (Nickel) ≤ 1.0 ppm
Pb (Lead) ≤ 0.5 ppm
Sb (Antimony) ≤ 5 ppm
Zn (Zinc) ≤ 2 ppm
Matter precipitated by ammonia passes test
Volatile acids (as Acetic acid) ≤ 10 ppm
Substances reducing potassium permanganate (as O) ≤ 10 ppm
Insoluble matter ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR