Perchloric acid 60%, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.00518.1001 1.00518.1016 1.00518.2501 1.00518.2514
CA1.00518.1001EA 405 CAD
CA1.00518.1001 CA1.00518.1016 CA1.00518.2501 CA1.00518.2514
Perchloric acid 60%, EMSURE® ACS for analysis, Supelco®
Perchloric acid
Formula: HClO₄
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112.2 °C
Density: 1.53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ag (Silver) ≤ 0.1 ppm
Al (Aluminium) ≤ 0.05 ppm
As (Arsenic) ≤ 0.05 ppm
Assay (acidimetric) 60.0 - 62.0 %
Ba (Barium) ≤ 0.02 ppm
Be (Beryllium) ≤ 0.02 ppm
Bi (Bismuth) ≤ 0.1 ppm
Ca (Calcium) ≤ 0.5 ppm
Cd (Cadmium) ≤ 0.05 ppm
Chlorate (ClO₃) ≤ 10 ppm
Chloride (Cl) ≤ 3 ppm
Co (Cobalt) ≤ 0.05 ppm
Color ≤ 10 Hazen
Cu (Copper) ≤ 0.1 ppm
Fe (Iron) ≤ 1.0 ppm
Free chlorine (Cl) ≤ 0.5 ppm
Ge (Germanium) ≤ 0.05 ppm
Heavy metals (as Pb) ≤ 1 ppm
K (Potassium) ≤ 0.1 ppm
Li (Lithium) ≤ 0.02 ppm
Mg (Magnesium) ≤ 0.5 ppm
Mn (Manganese) ≤ 0.02 ppm
Mo (Molybdenum) ≤ 0.05 ppm
Na (Sodium) ≤ 0.5 ppm
Ni (Nickel) ≤ 0.1 ppm
Pb (Lead) ≤ 0.05 ppm
Phosphate and Silicate (as SiO₂) ≤ 5 ppm
Residue on ignition (as sulfate) ≤ 30 ppm
Sr (Strontium) ≤ 0.02 ppm
Sulfate (SO₄) ≤ 10 ppm
Ti (Titanium) ≤ 0.1 ppm
Tl (Thallium) ≤ 0.05 ppm
Total nitrogen (N) ≤ 10 ppm
V (Vanadium) ≤ 0.05 ppm
Zn (Zinc) ≤ 0.1 ppm
Zr (Zirconium) ≤ 0.1 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR