Perchloric acid ≥70%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.00514.1000
1.00514.1006
CA1.00514.1000EA
854
CAD
CA1.00514.1000
CA1.00514.1006
Perchloric acid ≥70%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Perchloric acid
Formula:
HClO₄ Boiling Pt: 160…181 °C (1013 hPa) Melting Pt: –112.2 °C Density: 1.53…1.768 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 1873 ADR: 5.1,I |
|
Specification Test Results
Ag (Silver) | ≤ 0.1 ppm |
Al (Aluminium) | ≤ 0.05 ppm |
As (Arsenic) | ≤ 0.05 ppm |
Assay (acidimetric) | 70.0 - 72.0 % |
Ba (Barium) | ≤ 0.02 ppm |
Be (Beryllium) | ≤ 0.02 ppm |
Bi (Bismuth) | ≤ 0.1 ppm |
Ca (Calcium) | ≤ 0.5 ppm |
Cd (Cadmium) | ≤ 0.05 ppm |
Chlorate (ClO₃) | ≤ 10 ppm |
Chloride (Cl) | ≤ 3 ppm |
Co (Cobalt) | ≤ 0.05 ppm |
Color | ≤ 10 Hazen |
Cu (Copper) | ≤ 0.1 ppm |
Density (d 20/20) |
approx. 1.7 Conform to ACS, ISO, Reag.Ph Eur. |
Fe (Iron) | ≤ 1.0 ppm |
Free chlorine | ≤ 0.5 ppm |
Ge (Germanium) | ≤ 0.05 ppm |
Heavy metals (as Pb) | ≤ 1 ppm |
Hg (Mercury) | ≤ 0.005 ppm |
Identity | passes test |
K (Potassium) | ≤ 0.1 ppm |
Li (Lithium) | ≤ 0.02 ppm |
Mg (Magnesium) | ≤ 0.5 ppm |
Mn (Manganese) | ≤ 0.02 ppm |
Mo (Molybdenum) | ≤ 0.05 ppm |
Na (Sodium) | ≤ 0.5 ppm |
Ni (Nickel) | ≤ 0.1 ppm |
Pb (Lead) | ≤ 0.05 ppm |
Phosphate and Silicate (as SiO₂) | ≤ 5 ppm |
Residue on ignition (as sulfate) | ≤ 30 ppm |
Sr (Strontium) | ≤ 0.02 ppm |
Sulfate (SO₄) | ≤ 10 ppm |
Ti (Titanium) | ≤ 0.1 ppm |
Tl (Thallium) | ≤ 0.05 ppm |
Total nitrogen (N) | ≤ 10 ppm |
V (Vanadium) | ≤ 0.05 ppm |
Zn (Zinc) | ≤ 0.1 ppm |
Zr (Zirconium) | ≤ 0.1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...