Oxalic acid dihydrate 99.5-102.0% (by manganometry), EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Ethanedioic acid dihydrate
1.00495.0100
1.00495.0500
1.00495.1000
1.00495.9025
CA1.00495.0100EA
173
CAD
CA1.00495.0100
CA1.00495.0500
CA1.00495.1000
CA1.00495.9025
Oxalic acid dihydrate 99.5-102.0% (by manganometry), EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis, Supelco®
Oxalic acid dihydrate
Formula:
HO₂CCO₂H·2H₂O Boiling Pt: 149…160 °C (1013 hPa) (dec.) Melting Pt: 98…100 °C Density: 1.65 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149102 CAS Number: 6153-56-6 EINECS: 205-634-3 UN: 3261 ADR: 8,III REACH: 01-2119534576-33 Merck Index: 13,06980 |
|
Specification Test Results
Solubility | 102 g/l (20 °C) |
Melting Point | 101 °C |
Molar Mass | 126.07 g/mol |
Boiling Point | 149 - 160 °C (decomposition) |
Vapor Pressure | 21 hPa (50 °C) |
Flash Point | 157 °C (decomposition) |
Density | 1.65 g/cm3 (20 °C) |
pH | 1 (10 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...