Hydrofluoric acid 40%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.00338.2500
1.00338.2500
1.00338.1000
1.00338.1000
1.00338.0500
1.00338.0500
CA1.00338.2500CS
4189.66
CAD
EM1.00338.1000
EM1.00338.2500
CA1.00338.2500
CA1.00338.1000
EM1.00338.0500
CA1.00338.0500
Hydrofluoric acid 40%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Hydrofluoric acid
Formula:
HF MW: 20.01 g/mol |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 UN: 1790 ADR: 8,II |
Specification Test Results
Ag (Silver) | ≤ 0.02 ppm |
Al (Aluminium) | ≤ 0.05 ppm |
Arsenic and Antimony (as As) | ≤ 0.03 ppm |
Assay (acidimetric) | 40.0 - 42.0 % |
Ba (Barium) | ≤ 0.01 ppm |
Be (Beryllium) | ≤ 0.02 ppm |
Bi (Bismuth) | ≤ 0.02 ppm |
Ca (Calcium) | ≤ 0.2 ppm |
Cd (Cadmium) | ≤ 0.01 ppm |
Chloride (Cl) | ≤ 1 ppm |
Co (Cobalt) | ≤ 0.02 ppm |
Color | ≤ 10 Hazen |
Cr (Chromium) | ≤ 0.01 ppm |
Cu (Copper) | ≤ 0.02 ppm |
Fe (Iron) | ≤ 0.1 ppm |
Ge (Germanium) | ≤ 0.02 ppm |
Heavy metals (as Pb) | ≤ 0.5 ppm |
Hexafluorosilicate (SiF₆) | ≤ 50 ppm |
K (Potassium) | ≤ 0.1 ppm |
Li (Lithium) | ≤ 0.01 ppm |
Mg (Magnesium) | ≤ 0.1 ppm |
Mn (Manganese) | ≤ 0.03 ppm |
Mo (Molybdenum) | ≤ 0.02 ppm |
Na (Sodium) | ≤ 0.2 ppm |
Ni (Nickel) | ≤ 0.02 ppm |
Pb (Lead) | ≤ 0.02 ppm |
Phosphate (PO₄) | ≤ 0.5 ppm |
Residue on ignition (as sulfate) | ≤ 5 ppm |
Sr (Strontium) | ≤ 0.02 ppm |
Sulfate and sulphite (as SO₄) | ≤ 5 ppm |
Ti (Titanium) | ≤ 0.02 ppm |
Tl (Thallium) | ≤ 0.02 ppm |
V (Vanadium) | ≤ 0.02 ppm |
Zn (Zinc) | ≤ 0.05 ppm |
Zr (Zirconium) | ≤ 0.02 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...