Hydrofluoric acid 38 - 40%, extra_pure, Supelco®
Supplier: MilliporeSigma
|
Danger
|
MQ200
1.00337.1000
1.00337.2500
CA1.00337.1000CS
1040
CAD
CA1.00337.1000
CA1.00337.2500
Hydrofluoric acid 38 - 40%, extra_pure, Supelco®
Hydrofluoric acid
Formula:
HF MW: 20.01 Storage Temperature: Ambient |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 EINECS: 231-634-8 UN: 1790 ADR: 8 II |
Specification Test Results
Assay (acidimetric) | 38 - 40 % |
Chloride (Cl) | ≤ 0.002 % |
Fe (Iron) | ≤ 0.001 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
Hexafluorosilicate (SiF₆) | ≤ 0.01 % |
Residue on ignition (as sulfate) | ≤ 0.002 % |
Sulfate (SO₄) | ≤ 0.001 % |
Sulfite (SO₃) | ≤ 0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...