Hydrochloric acid 32% (by alkalimetry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00319.1000 1.00319.1011 1.00319.2500 1.00319.2511 1.00319.9025
CA1.00319.1000CS 418 CAD
CA1.00319.1000 CA1.00319.1011 CA1.00319.2500 CA1.00319.2511 CA1.00319.9025
Hydrochloric acid 32% (by alkalimetry), EMSURE® for analysis, Supelco®
Hydrochloric acid
Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.16 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -40 °C
Boiling Point 83 - 90 °C
Vapor Pressure 21.3 hPa (20 °C)
Density 1.16 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR