Hydrochloric acid fuming 37%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.00317.1000 1.00317.2500 1.00317.2510 1.00317.9026
CA1.00317.1000CS 1030 CAD
CA1.00317.1000 CA1.00317.2500 CA1.00317.2510 CA1.00317.9026
Hydrochloric acid fuming 37%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Hydrochloric acid
Formula: HCl
Boiling Pt: >100 °C (lit.)
Melting Pt: –30 °C
Density: 1.19 g/cm3 (20 °C), 1.2 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Ag (Silver) ≤ 0.020 ppm
Al (Aluminium) ≤ 0.050 ppm
Appearance passes test
Appearance of solution passes test
As (Arsenic) ≤ 0.010 ppm
Assay (alkalimetric) 37.0 - 38.0 %
Au (Gold) ≤ 0.050 ppm
B (Boron) ≤ 0.100 ppm
Ba (Barium) ≤ 0.010 ppm
Be (Beryllium) ≤ 0.010 ppm
Bi (Bismuth) ≤ 0.050 ppm
Bromide (Br) ≤ 50 ppm
Ca (Calcium) ≤ 0.300 ppm
Cd (Cadmium) ≤ 0.010 ppm
Co (Cobalt) ≤ 0.010 ppm
Color ≤ 10 Hazen
Cr (Chromium) ≤ 0.010 ppm
Cu (Copper) ≤ 0.010 ppm
Expiry date see product label
Fe (Iron) ≤ 0.100 ppm
Free chlorine (Cl) ≤ 1 ppm
Ga (Gallium) ≤ 0.050 ppm
Ge (Germanium) ≤ 0.020 ppm
Heavy metals (as Pb) ≤ 1 ppm
Hg (Mercury) ≤ 0.010 ppm
Identity passes test
K (Potassium) ≤ 0.100 ppm
Li (Lithium) ≤ 0.010 ppm
Mg (Magnesium) ≤ 0.050 ppm
Mn (Manganese) ≤ 0.010 ppm
Mo (Molybdenum) ≤ 0.010 ppm
Na (Sodium) ≤ 0.300 ppm
NH₄ (Ammonium) ≤ 1 ppm
Ni (Nickel) ≤ 0.020 ppm
Non volatile matter ≤ 10 ppm
Pb (Lead) ≤ 0.010 ppm
Phosphate (PO₄) ≤ 0.5 ppm
Pt (Platinum) ≤ 0.100 ppm
Residue on ignition (as SO₄) ≤ 3 ppm
Sn (Tin) ≤ 0.050 ppm
Sr (Strontium) ≤ 0.010 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Sulfite (SO₃) ≤ 0.5 ppm
Ti (Titanium) ≤ 0.020 ppm
Tl (Thallium) ≤ 0.020 ppm
V (Vanadium) ≤ 0.010 ppm
Zn (Zinc) ≤ 0.050 ppm
Zr (Zirconium) ≤ 0.020 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR