Hydrobromic acid 47%, Extra Pure, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.00304.0500
1.00304.2500
1.00304.9020
CA1.00304.0500CS
1278
CAD
CA1.00304.0500
CA1.00304.2500
CA1.00304.9020
Hydrobromic acid 47%, Extra Pure, Supelco®
Hydrogen bromide
Formula:
HBr Density: 0.79 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011323 CAS Number: 10035-10-6 EINECS: 231-595-7 UN: 1788 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -11 °C |
Boiling Point | 126 °C (1013 hPa) |
Vapor Pressure | 10.6 hPa (20 °C) |
Density | 1.49 g/cm3 (20 °C) |
pH | (H2O, 20 °C) strongly acid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...