Phenol 99.0-100.5% (by bromometric analysis) Reag. Ph. Eur., ACS, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

1.00206.0250 1.00206.1000 1.00206.9025
CA1.00206.0250CS 611 CAD
CA1.00206.0250 CA1.00206.1000 CA1.00206.9025
Phenol 99.0-100.5% (by bromometric analysis) Reag. Ph. Eur., ACS, GR for analysis, Supelco®
Phenol

1.00206.0250 is offered as an alternative to PX0510-1. Different Pack Size, Same Specifications.
1.00206.1000 replaces and is the same as PX0510-2. Part # Change only, Same Pack Size, Same Specifications.

Formula: C₆H₅OH
Boiling Pt: 182 °C (lit.)
Melting Pt: 40…42 °C (lit.)
Density: 1.071 g/ml (25°C) (lit.)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
REACH: 01-2119471329-32
Merck Index: 12,07390

Order Now

Specification Test Results

Solubility 84 g/l (20 °C)
Melting Point 40.8 °C
Molar Mass 94.11 g/mol
Bulk Density 620 kg/m3
Boiling Point 181.8 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 81 °C
Explosion Limit 1.3 - 9.5 %(V)
Density 1.06 g/cm3 (20 °C)
pH 5 (50 g/l, H2O, 20 °C)
Ignition Point 595 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR