Amidosulfonic acid, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Sulfamic acid, Sulfamidic acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid, Aminosulfonic acid, Amidosulfonic acid, Amidosulfuric Acid, Amidosulfuric acid

1.00103.0100 1.00103.0250
CA1.00103.0100EA 141 CAD
CA1.00103.0100 CA1.00103.0250
Amidosulfonic acid, EMSURE® for analysis, Supelco®
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97.09 g/mol
Boiling Pt: 247
Melting Pt: 205
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8 III
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 213 g/l (20 °C)
Melting Point 205 °C (decomposition)
Molar Mass 97.08 g/mol
Bulk Density 600 kg/m3
Density 2.13 g/cm3 (20 °C)
pH 1.2 (10 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR