Acetic acid glacial, anhydrous ≥99.8% (by alkalimetry), EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

M-Clarity™ quality level = MQ300

1.00063.1000 1.00063.1011 1.00063.2500 1.00063.2510 1.00063.2511 1.00063.9026
CA1.00063.1000EA 751 CAD
CA1.00063.1000 CA1.00063.1011 CA1.00063.2500 CA1.00063.2510 CA1.00063.2511 CA1.00063.9026
Acetic acid glacial, anhydrous ≥99.8% (by alkalimetry), EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 117…118 °C (lit.)
Melting Pt: 16.2 °C (lit.)
Density: 1.049 g/ml (25°C) (lit.)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8.3,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric) ≥ 99.8 %
Identity passes test
Color ≤ 10 Hazen
Acetaldehyde ≤ 2 ppm
Acetic anhydride ≤ 100 ppm
Titratable base ≤ 0.0004 meq/g
Solidification temperature ≥ 16.3 °C
Chloride (Cl) ≤ 0.4 ppm
Phosphate (PO₄) ≤ 0.4 ppm
Heavy metals (as Pb) ≤ 0.5 ppm
Sulfate (SO₄) ≤ 0.4 ppm
Ag (Silver) ≤ 0.005 ppm
Al (Aluminium) ≤ 0.020 ppm
As (Arsenic) ≤ 0.010 ppm
Au (Gold) ≤ 0.010 ppm
B (Boron) ≤ 0.100 ppm
Ba (Barium) ≤ 0.010 ppm
Be (Beryllium) ≤ 0.005 ppm
Bi (Bismuth) ≤ 0.050 ppm
Ca (Calcium) ≤ 0.100 ppm
Cd (Cadmium) ≤ 0.020 ppm
Co (Cobalt) ≤ 0.010 ppm
Cr (Chromium) ≤ 0.020 ppm
Cu (Copper) ≤ 0.010 ppm
Fe (Iron) ≤ 0.050 ppm
Ga (Gallium) ≤ 0.050 ppm
Ge (Germanium) ≤ 0.020 ppm
Hg (Mercury) ≤ 0.005 ppm
In (Indium) ≤ 0.050 ppm
K (Potassium) ≤ 0.100 ppm
Li (Lithium) ≤ 0.010 ppm
Mg (Magnesium) ≤ 0.050 ppm
Mn (Manganese) ≤ 0.010 ppm
Mo (Molybdenum) ≤ 0.010 ppm
N/A (Sodium) ≤ 0.200 ppm
Ni (Nickel) ≤ 0.020 ppm
Pb (Lead) ≤ 0.010 ppm
Pt (Platinum) ≤ 0.100 ppm
Sn (Tin) ≤ 0.050 ppm
Sr (Strontium) ≤ 0.010 ppm
Ti (Titanium) ≤ 0.050 ppm
Tl (Thallium) ≤ 0.020 ppm
V (VaN/Adium) ≤ 0.010 ppm
Zn (Zinc) ≤ 0.030 ppm
Zr (Zirconium) ≤ 0.050 ppm
Substances reducing potassium dichromate passes test
Substances reducing potassium permangaN/Ate ≤ 20 ppm
Evaporation residue ≤ 5 ppm
Water ≤ 0.2
Dilution test passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR