GelCode™ Blue Safe Protein Stain, Thermo Scientific

Supplier: Thermo Scientific

24594 24596
CAPI24594EA 308.76 CAD
CAPI24594 CAPI24596
GelCode™ Blue Safe Protein Stain, Thermo Scientific
Stains Electrophoresis Stains

GelCode Blue Safe Protein Stain is a fast and sensitive coomassie-dye reagent that provides one-step ultrasensitive detection of proteins in polyacrylamide gels and is formulated to avoid hazardous shipping.


  • Sensitivity—detect down to 9 ng of protein per band in 15 minutes
  • Safety—non-corrosive to skin, non-flammable, and safe to ship and store until activated
  • Additional reagents—water washes only; no acid-fixative or methanol destain solutions required
  • Compatibility—suitable for staining 1D and 2D polyacrylamide gels and nitrocellulose and PVDF membranes (multiple protocols provided in the instructions)
  • Applications—qualitative visualization, quantitative densitometry, and downstream excision and analysis by mass spectrometry
  • Easy to use—add and dissolve activator crystals before first use, then simply add reagent to stain gels
  • Stable—store non-activated or activated stain at room temperature for up to one year


For proper protein staining, the Activator Crystals must be dissolved in the staining reagent before use. Add the entire contents of the Activator Crystals bottle into the GelCode Blue Safe Protein Stain bottle. Dissolve the crystals by mixing end-over-end for 1 minute, and then allow it to remain at room temperature for 5 minutes before use. Store the activated stain at room temperature.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR