Tetrahydrofuran, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Filtered through a 0.2 µm filter, packaged under nitrogen.

CABDH28559.400 BDH28559.100
CABDH28559.400CS 10134.07 CAD
CABDH28559.400 BDH28559.100
Tetrahydrofuran, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Tetrahydrofuran

Not stabilized.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) (Corrected for Water) Min. 99.70%
Water (by KF coulometry) Max. 0.1000%
Residue after evaporation Max. 0.0005%
Acidity Max. 0.0005 meq/g
Transmittance at 230 nm Min. 40%
Transmittance at 240 nm Min. 60%
Transmittance at 250 nm Min. 70%
Transmittance at 260 nm Min. 80%
Transmittance at 280 nm Min. 96%
Transmittance at 300 nm Min. 98%
Transmittance (300 nm) Min. 98 %
Conforms to BDH 15247 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR