N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH83634.100
BDH83634.400
BDH83634.100CS
949.81
CAD
BDH83634.100
BDH83634.400
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
N,N-Dimethylformamide
Formula:
HCON(CH₃)₂ Boiling Pt: 153 °C (1013 hPa) Melting Pt: –61 °C Density: 0.949 g/cm³ (20 °C) Flash Pt: 57.5 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 13,03269 |
|
Specification Test Results
Assay (by GC-FID) | Min. 99.9 % |
Water (by KF coulometry) | Max. 0.02 % |
Nonvolatile Residue | Max. 0.0005 % |
Acidity | Max. 0.0005 meq/g |
Alkalinity | Max. 0.0002 meq/g |
Transmittance (270 nm) | Min. 10 % |
Transmittance (275 nm) | Min. 50 % |
Transmittance (290 nm) | Min. 80 % |
Transmittance (300 nm) | Min. 90 % |
Transmittance (330 nm) | Min. 98 % |
Conforms to BDH 15297 | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...