2-Propanol ≥99.8% (by GC), HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

BDH20880.400 BDH20880.100 BDH20880.100E
BDH20880.400CS 1438.35 CAD
BDH20880.400 BDH20880.100 BDH20880.100E
2-Propanol ≥99.8% (by GC), HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
2-Propanol

From the exclusive family of high-quality, competitively priced BDH Chemicals — high purity solvents for HPLC applications.


  • Designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Filtered down to 0.2 micron level and bottled under Nitrogen
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally-friendly corrugated/pulp case inserts that are fully recyclable

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Water Max. 0.1 %
Evaporation residue 5 ppm
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Absorbance (220 nm) Max. 0.3
Absorbance (230 nm) Max. 0.1
Absorbance (250 nm) Max. 0.02
Transmittance (220 nm) Min. 50 %
Transmittance (230 nm) Min. 80 %
Transmittance (250 nm) Min. 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR