Buckets with Casters, 36 L, Perfex
Supplier: Perfex
30-36R 30-36W 30-36B
89096-014EA 271.13 CAD
89096-014 89096-016 89096-018
Buckets with Casters, 36 L, Perfex
Buckets
High temperature resistant polypropylene bucket
  • Non-porous, super smooth surface prevents microbial growth
  • Embossed graduations in English and Metric for accurate measure
  • Lightweight and durable, includes 3-inch twin-wheel casters with one locking caster
  • Easily accessible brake pedal allows parking on uneven floors
Compatible with gamma, ETO and autoclave sterilization – up to 250°F (121°C) for 30 minutes.