TruCLEAN Deluxe Disinfection System, Perfex
Supplier: Perfex
30-3R 30-3W 30-3B
89203-552EA 721.19 CAD
89203-552 89203-554 89203-556
TruCLEAN Deluxe Disinfection System, Perfex
Mops
Designed for small area cleaning and disinfecting
Compatible with gamma, ETO and autoclave sterilization – up to 250°F (121°C) for 30 minutes.
Includes 36-liter bucket with 3” casters, waste containment bucket, stainless steel sieves (one for each bucket), stainless steel mop frame and polymer adjustable mop handle. Captures and isolates soiled contaminants from adulterating cleaning and disinfecting agents. Embossed graduations in English and Metric for accurate measure. Quickly and effectively disinfect surfaces in surgical areas and controlled contamination zones.