n-Hexane ≥97%, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Filtered through a 0.2 µm filter, packaged under nitrogen.

BDH24575.400 CABDH24575.400 BDH24575.100E
BDH24575.400CS 1194.52 CAD
BDH24575.400 CABDH24575.400 BDH24575.100E
n-Hexane ≥97%, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
n-Hexane

From the exclusive family of high-quality, competitively priced BDH Chemicals — high purity solvents for HPLC applications.


  • Designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Filtered down to 0.2 micron level and bottled under Nitrogen
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally-friendly corrugated/pulp case inserts that are fully recyclable

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 97 %
Water Max. 0.005 %
Non-volatile residue Max. 0.0005 %
Acidity Max. 0.0003 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (210 nm) Min. 50 %
Transmittance (220 nm) Min. 80 %
Transmittance (245 nm) Min. 98 %
Conforms to BDH 15249 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR