Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Methylene chloride

Stabilization: Stabilised with 2-methyl-2-butene (iso-amylene) 20 ppm
Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRCBDH23373.4 BDHVBDH23373.100E
BDH23373.400CS 1030.72 CAD
BDH23373.400 BDH23373.100E
Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Dichloromethane

From the exclusive family of high-quality, competitively priced BDH Chemicals — high purity solvents for HPLC applications.


  • Designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Filtered down to 0.2 micron level and bottled under Nitrogen
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally-friendly corrugated/pulp case inserts that are fully recyclable


Stabilized with ±0.002% 2-methyl-2-butene.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Water Max. 0.05 %
Non-volatile residue Max. 0.001 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (240 nm) Min. 50 %
Transmittance (245 nm) Min. 80 %
Transmittance (260 nm) Min. 98 %
Conforms to BDH 15245 Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR